ContactUsLinkComponent
Gewindeoberteil Service temperature range of PFA is -328F to 500F (-200C to 260C) PFA non-breakable Threaded top for long term storage Fits most standard centrifuge rotors RCF up to 5,000 g 1 * 1 ST
Catalog # SAVI210-050-30
Supplier:  SAVILLEX
Gewindeoberteil Service temperature range of PFA is -328F to 500F (-200C to 260C) PFA non-breakable Threaded top for long term storage Fits most standard centrifuge rotors RCF up to 5,000 g 1 * 1 ST
Catalog # SAVI210-050-30
Supplier:  SAVILLEX
Frequently Bought Together