ContactUsLinkComponent
Cooling unit fast MTR 115v/60Hz, high cooling capacity up to -40C, Manual defrost, Extremely silent due to special valves, Self opt imizing controller for temperature accuracy 1 * 1 items
Catalog # SLEE10110002
Supplier:  Slee
Cooling unit fast MTR 115v/60Hz, high cooling capacity up to -40C, Manual defrost, Extremely silent due to special valves, Self opt imizing controller for temperature accuracy 1 * 1 items
Catalog # SLEE10110002
Supplier:  Slee
Frequently Bought Together