ContactUsLinkComponent
n6-carbobenzyloxy-n2,n2-bis(carboxymethyl)-l-lysine is used in the synthesis of nickel-chelating fluorinated lipids for protein mono layer crystallizations. 1 * 1 g
Catalog # GENO786-949
Supplier:  G-Biosciences
n6-carbobenzyloxy-n2,n2-bis(carboxymethyl)-l-lysine is used in the synthesis of nickel-chelating fluorinated lipids for protein mono layer crystallizations. 1 * 1 g
Catalog # GENO786-949
Supplier:  G-Biosciences
Frequently Bought Together