ContactUsLinkComponent
ym agar is used to cultivate yeasts, molds and other aciduric organisms. consists of 5g/l enzymatic digest of gelatin, 3g/l yeast e xtract, 3g/l malt extract, 10g/l dextrose, 20g/l agar at a final ph of 6.2. 1 * 125 mL
Katalognr. GENOBTNM-0095
Supplier:  G-Biosciences
ym agar is used to cultivate yeasts, molds and other aciduric organisms. consists of 5g/l enzymatic digest of gelatin, 3g/l yeast e xtract, 3g/l malt extract, 10g/l dextrose, 20g/l agar at a final ph of 6.2. 1 * 125 mL
Katalognr. GENOBTNM-0095
Supplier:  G-Biosciences
Frequently Bought Together