ContactUsLinkComponent
Microscope slides with reaction wells, 76x26 mm, thickness 1 mm, white glass, edges ground at 90, with frosted end, 10 numbered, wells of 6 mm diameter, white epoxy resin mask, surface treated with silane 1 * 50 items
Catalog # MARI1235292
Supplier:  MARIENFELD
Microscope slides with reaction wells, 76x26 mm, thickness 1 mm, white glass, edges ground at 90, with frosted end, 10 numbered, wells of 6 mm diameter, white epoxy resin mask, surface treated with silane 1 * 50 items
Catalog # MARI1235292
Supplier:  MARIENFELD
Frequently Bought Together