ContactUsLinkComponent
Safety-can 1/2l with screw cap 1inch, flame trap and pressure control valve 1 * 1 items
Catalog # ROET0HK
Safety-can 1/2l with screw cap 1inch, flame trap and pressure control valve 1 * 1 items
Catalog # ROET0HK
Frequently Bought Together