Orders LinkContactUsLinkComponent
Ketone Control Solution for GK Dual Meter, Contents: 2ml Control Solution 0, 2ml Control Solution 1, 2ml Control Solution 2, Package insert 1 * 1 items
Ketone Control Solution for GK Dual Meter, Contents: 2ml Control Solution 0, 2ml Control Solution 1, 2ml Control Solution 2, Package insert 1 * 1 items
Catalog # SUREGKKETCK
Supplier:   SURESCREEN
Ketone Control Solution for GK Dual Meter, Contents: 2ml Control Solution 0, 2ml Control Solution 1, 2ml Control Solution 2, Package insert 1 * 1 items
Catalog # SUREGKKETCK
Supplier:  SURESCREEN
Ketone Control Solution for GK Dual Meter, Contents: 2ml Control Solution 0, 2ml Control Solution 1, 2ml Control Solution 2, Package insert 1 * 1 items
Price
SFr. 19.10
Log in for availability
1 * 1 ST