ContactUsLinkComponent
HPLC guard column EC 4/3 NUCLEOSIL 100-10 SB lenght: 4 mm, ID: 3 mm pack of 3 - require Column Protection System guard column holde r (REF 718966) - for anayltical columns with 3, 4 and 4.6 mm ID 1 * 3 items
Catalog # MANA721164.30
Supplier:  MACHEREY-NAGEL
HPLC guard column EC 4/3 NUCLEOSIL 100-10 SB lenght: 4 mm, ID: 3 mm pack of 3 - require Column Protection System guard column holde r (REF 718966) - for anayltical columns with 3, 4 and 4.6 mm ID 1 * 3 items
Catalog # MANA721164.30
Supplier:  MACHEREY-NAGEL
Frequently Bought Together