Boat-combustion Quartz glass in application temperatures of up to 1050°C (maximum 1600°C), with handle, translucent, high chemical resistance and purity. length 78mm, width 17mm, height 11mm. The durability or operating cycles of our Boat-combustion 1 * 1 items
Catalog # TSLA73380
Supplier: TECHNICAL QUARTZ SOLUTIONS
Frequently Bought Together