Sterile goods bascket 1/2 StU 575 x 280 x 135 mmn, stainless steel electropolished, with handle opening, stackable 1 * 1 items
Catalog # HUPF7500016
Supplier: HUPFER METALLWERKE
Sterile goods bascket 1/2 StU 575 x 280 x 135 mmn, stainless steel electropolished, with handle opening, stackable 1 * 1 items
Catalog # HUPF7500016
Supplier: HUPFER METALLWERKE
Frequently Bought Together
Reducing screw thread adaptor couplings, BOLA
Supplier: Bohlender
VWR®, Measuring Beakers with Handle, PP
Supplier: VWR Collection
VWR®, Microtube Storage Boxes
Supplier: VWR Collection
VWR®, Sleeves for Standard Ground Joints
Supplier: VWR Collection
Multiple distributors with stopcock for bottles with GL 45 thread, BOLA
Supplier: Bohlender
Multiple distributors for bottles with GL 45 thread, BOLA
Supplier: Bohlender
Pipette tips
Supplier: Sartorius
Buckets, Round, PP
Supplier: BURKLE
Test tube racks, Unwire™, Nalgene®
Supplier: Thermo Fisher Scientific
Surface protection sheets, BOLA Sheets PTFE or FEP
Supplier: Bohlender
HT (high temperature) laboratory connectors with GL thread
Supplier: Bohlender
ep Dualfilter T.I.P.S.® SealMax® Racks Self-Sealing Pipette Tips
Supplier: EPPENDORF
BASKET STERILE GOODS 575X280MM 1 * 1 items
Supplier: HUPFER METALLWERKE
Serum separator tubes, SST™, BD Vacutainer®
Supplier: BD Medical
MONITORING SHELF 575/390/155 SS E-POLISC 1 * 1 items
Supplier: HUPFER METALLWERKE
epT.I.P.S.® Reloads, Pipette Tips
Supplier: EPPENDORF