Closures
Screw cap, completed with dispensing- insert, RD 25, White, PP
Catalog # RIXI8-2031-25-W
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
Product Details & Documents
Specifications
- Thread:RD 25
- Colour:White
- Description:Screw cap, completed with dispensing- insert
- Material:PP
- Seq:27