Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Accessories

Accessories

Delivery jet assembly, 90 mm, for 1 ml dispenser
Catalog # SOCO1.525.090
Supplier:   Socorex Isba
Accessories
Catalog # SOCO1.525.090
Supplier:  Socorex Isba
Accessories
Accessories

Delivery jet assembly, 90 mm, for 1 ml dispenser

Price
SFr. 75.10
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Delivery jet assembly, 90 mm, for 1 ml dispenser
  • Seq:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.