ContactUsLinkComponent
Replacement feed tubing, PTFE, cut 300 mm, for CalibrexTM 520/525/530 up to 10 ml 1 * 1 items
Catalog # SOCO511.707
Supplier:  Socorex Isba
Replacement feed tubing, PTFE, cut 300 mm, for CalibrexTM 520/525/530 up to 10 ml 1 * 1 items
Catalog # SOCO511.707
Supplier:  Socorex Isba
Frequently Bought Together