ContactUsLinkComponent
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 40 x 52 cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1291
Stericlin® sterilisation pouches, flat, non-woven 60g/m² with see-through film, with indicators for STEAM, EO and FORM, 40 x 52 cm, DIN EN ISO 11607 1 * 250 items
Catalog # 129-1291