ContactUsLinkComponent
Mustimeter with traceable calibration certificate range : 0.980-1.130 unit : g/ml div. : 0.001 lenght mm : 350 accuracy : ± 0.001 calibration temp. : 20°C thermometer in body : 0+40°C : 1°C 1 * 1 items
Codice catalogo ALLA2001TC1720EUCA
Nome del fornitore:  Alla France
Mustimeter with traceable calibration certificate range : 0.980-1.130 unit : g/ml div. : 0.001 lenght mm : 350 accuracy : ± 0.001 calibration temp. : 20°C thermometer in body : 0+40°C : 1°C 1 * 1 items
Codice catalogo ALLA2001TC1720EUCA
Nome del fornitore:  Alla France
Frequently Bought Together