Lysosome-associated membrane proteins (LAMP) are glycosylated type I membrane proteins that play a role in the biogenesis of the pig ment melanin. LAMP-1 (also designated CD107A) and LAMP-2 (also designated CD107B) are involved in a variety of functi 1 * 100 µG
Catalog # BWRLBS61513
Supplier: Bioworld Technology
Frequently Bought Together