ContactUsLinkComponent
Mµlti OneTouch Standard Tips 1 - 3 µl, sterile 10 Cell Racks of 96 Tips Filter Tips made of crystal-clear polypropylene and flexible TPE, manufactured with two-component injection moulding technique. Certified free from RNase, DNase, pyrogens, human 1 * 960 items
Catalog # SORE78490
Mµlti OneTouch Standard Tips 1 - 3 µl, sterile 10 Cell Racks of 96 Tips Filter Tips made of crystal-clear polypropylene and flexible TPE, manufactured with two-component injection moulding technique. Certified free from RNase, DNase, pyrogens, human 1 * 960 items
Catalog # SORE78490
Frequently Bought Together