LISTERIA FRASER BROTH - 20 x 10 ML 1 * 10 mL
Catalog # BLIF551596
Supplier: BIOLIFE ITALIANA
Frequently Bought Together
VWR® Culture Media, Ready to Use, in Bottles
Supplier: VWR Chemicals
Media supplements for dehydrated culture media
Supplier: Merck
VWR® Ready to Use Media Plates (90 mm), Irradiated
Supplier: VWR Chemicals
VWR® Culture Media, Ready To Use, In Plates
Supplier: VWR Chemicals
Antibiotic assay discs, differentiation discs (DD)
Supplier: THERMO FISHER DIAGNOSTICS
SELECTIVE SUPPLEMENT MUP VIALS 1 * 10 items
Supplier: VWR Chemicals
LUBIOSCIENC QUICKBLUE PROTEIN STAI
Supplier: MARKET SOURCE PART PROCESS
VWR® Contact Plates for Surface and Air Control, Irradiated
Supplier: VWR Chemicals
Buffered NaCl Peptone Solution pH 7,0
Supplier: LIOFILCHEM
FASSIF BUFFER CONCENTRATE (6L) 250G 1 * 250 g
Supplier: MARKET SOURCE PART PROCESS
Ready Culture Media in Tubes
Supplier: LIOFILCHEM
Antibiotic assay discs
Supplier: LIOFILCHEM
Dehydrated Media
Supplier: THERMO FISHER DIAGNOSTICS
Pyrosol ® Buffer without pH indicator, 5 vials per pack, each vial contains 5.5 ml 1 * 5 items
Supplier: Associates of Cape Cod
TRYPTIC SOY AGAR, FOR MICROBIOLOGY 1 * 500 g
Supplier: Merck