AMMONIUM IONIC STRENGTH ADJUSTOR SOLN 1 * 500 mL
Catalog # YSIA400374
Supplier: YSI ENVIRONMENTAL
Frequently Bought Together
Sulfamethoxazole, Reference Standard
Supplier: USP
Supelco® PESTANAL® Organic Reference Standard, Phenmedipham
Supplier: Merck
Fenoxaprop-P (R-enantiomer) certified 1 * 0,1 g
Supplier: EHRENSTORFER
2-Butanol, Reference Standard
Supplier: USP
Reference standard for food analysis, phthalic acid, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Supelco® TraceCERT® Organic Reference Standard, Fenazaquin
Supplier: Merck
Pesticide reference standard, Endrin 10 µg/ml in acetonitrile, Dr Ehrenstorfer
Supplier: EHRENSTORFER
Fonofos certified 1 * 0,1 g
Supplier: EHRENSTORFER
Polyvinyl Alcohol, Reference Standard
Supplier: USP
TRULINE AMMONIUM LOW STANDARD 1 * 500 mL
Supplier: YSI ENVIRONMENTAL
FLORASULAM CERTIFIED 1 * 50 mg
Supplier: EHRENSTORFER
Nitrate standards for ISE calibration, Orion™
Supplier: Thermo Orion