ContactUsLinkComponent
2 Liter BPA free, Copolyester container, 4 programmable settings, LCD display, programmable speed 1.500-20.000 RPM, dual pulse function, dishwasher safe removable jar pad. Accessories like smaller 1.25 L glass container CAC32 and adapter AD3 1 * 1 items
Catalog # WARIMX1300XTXEE
Supplier:  WARING BLENDOR
2 Liter BPA free, Copolyester container, 4 programmable settings, LCD display, programmable speed 1.500-20.000 RPM, dual pulse function, dishwasher safe removable jar pad. Accessories like smaller 1.25 L glass container CAC32 and adapter AD3 1 * 1 items
Catalog # WARIMX1300XTXEE
Supplier:  WARING BLENDOR
Frequently Bought Together