ContactUsLinkComponent
Precision Milk Butyrometer % Fat: 0 - 3 - 6 division: 0.05 With a frosted label on the bulb, plain neck, without a stopper, for milk quantities of 10.73 - 11 ml, accuracy of scale ± 0.03%. 1 * 1 items
Catalog # GEAG03.0043
Precision Milk Butyrometer % Fat: 0 - 3 - 6 division: 0.05 With a frosted label on the bulb, plain neck, without a stopper, for milk quantities of 10.73 - 11 ml, accuracy of scale ± 0.03%. 1 * 1 items
Catalog # GEAG03.0043
Frequently Bought Together