ContactUsLinkComponent
INCLUDES 3 ML SYRINGE, TWO LUER TO 1/4-28 ADAPTERS. USE SYRINGE AND WHITE ADAPTER TO BACK-FLUSH THE CARRIER LINE OR TEE-LINE. USE SY RINGE AND RED ADAPTER TO FORWARD FLUSH THE CARRIER LINE, THE SIDE PORT OF THE TEE-LINE, AND TO FORWARD FLUSH THE PFA 1 * 1 items
  PERCN8121035
INCLUDES 3 ML SYRINGE, TWO LUER TO 1/4-28 ADAPTERS. USE SYRINGE AND WHITE ADAPTER TO BACK-FLUSH THE CARRIER LINE OR TEE-LINE. USE SY RINGE AND RED ADAPTER TO FORWARD FLUSH THE CARRIER LINE, THE SIDE PORT OF THE TEE-LINE, AND TO FORWARD FLUSH THE PFA 1 * 1 items
  PERCN8121035
Frequently Bought Together