Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Viscosity reference standards

Viscosity reference standards

Nominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 3,8 l, Hydrocarbons, 1
Catalog # PERCN5316025
Supplier:   PerkinElmer
Viscosity reference standards
Catalog # PERCN5316025
Supplier:  PerkinElmer
CAS Number:  
Viscosity reference standards
Viscosity reference standards

Nominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 3,8 l, Hydrocarbons, 1

CAS Number:  
Price
SFr. 1’370.00
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications

Specifications

  • Pk:
    1
  • Description:
    Nominal viscosity: 30 cSt at 40 °C and 5,3 cSt at 100 °C, 3,8 l
  • Type:
    Hydrocarbons

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.