CAP FOR HIGH PRESSURE 100 ML (100 BAR) DIGESTION VESSELS WITHOUT DPC PRESSURE SENSOR FOR TITAN MPS 8 POSITION MICROWAVE SAMPLE PREPA RATION SYSTEMS. INCLUDES VESSEL CAP MADE OF TFM WITH PRESSURE SENSOR FOR HIGH PRESSURE 100 ML (100 BAR) DIGESTION VES 1 * 1 items
Catalog # PERCN3133016
Supplier: PerkinElmer
Frequently Bought Together