ContactUsLinkComponent
EACH V-21+K WEAR METALS STANDARD 300 µG/G, HYDROCARBON OIL, 200 G INCLUDES THE FOLLOWING ELEMENTS: AG, AL, BA, B, CD, CA, CR, CU, FE , K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN CONCENTRATION: 300 µG/GMATRIX: HYDROCARBON OIL. VOLUME: 200 G. V-21 1 * 1 items
Codice catalogo PERCN9308364
Nome del fornitore:  PerkinElmer
EACH V-21+K WEAR METALS STANDARD 300 µG/G, HYDROCARBON OIL, 200 G INCLUDES THE FOLLOWING ELEMENTS: AG, AL, BA, B, CD, CA, CR, CU, FE , K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN CONCENTRATION: 300 µG/GMATRIX: HYDROCARBON OIL. VOLUME: 200 G. V-21 1 * 1 items
Codice catalogo PERCN9308364
Nome del fornitore:  PerkinElmer
Frequently Bought Together