ContactUsLinkComponent
Skap2 - Mouse, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 54353). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector, TR30021, included for free 1 * 1 KIT
Catalog # ORIGTL503013
Supplier:  OriGene
Skap2 - Mouse, 4 unique 29mer shRNA constructs in lentiviral GFP vector (Gene ID = 54353). 5µg purified plasmid DNA per construct Non-effective 29-mer scrambled shRNA cassette in pGFP-C-shLenti Vector, TR30021, included for free 1 * 1 KIT
Catalog # ORIGTL503013
Supplier:  OriGene
Frequently Bought Together