Sodium hydroxide pellets EMPROVE EXPERT Ph Eur,BP,JP,NF,ACS 1 * 1 kg
Catalog # 1.37020.1000
Supplier: MERCK PRODUCTION CHEMICALS
Sodium hydroxide pellets EMPROVE EXPERT Ph Eur,BP,JP,NF,ACS 1 * 1 kg
Catalog # 1.37020.1000
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 70% ≥70% (v/v), TechniSolv®
Supplier: VWR Chemicals
Eppendorf® Xplorer® Plus Electronic Single-Channel Pipettes
Supplier: EPPENDORF
Safety shoes, slip-on, cream
Supplier: Jallatte