Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Sump Pallets

Sump Pallets

Sump pallet, PE, 250 l with PE grating, load capacity 1000 kg, weight 25,7 kg, 1600×1200×180 mm
Catalog # CEMG10713
Supplier:   CEMO
Sump Pallets
Catalog # CEMG10713
Supplier:  CEMO
Sump Pallets
Sump Pallets

Sump pallet, PE, 250 l with PE grating, load capacity 1000 kg, weight 25,7 kg, 1600×1200×180 mm

Price
SFr. 821.00
Log in for availability
1 * 1 ST
Product Details & Documents

These small container/laboratory trays can be combined to cover a complete storage area and also can be used as storage/filling station. Trays can be used with or without perforated plate.

  • Made of polyethylene
  • With general construction inspection approval (application for admission)
  • Stackable and provides space-saving storage/transport
  • Fully recyclable and maintenance friendly
  • Rugged, removable PE grating
  • Can be connected to form a surface protection system

Stable PE access ramp with an integrated fixing bolt for connection to the PE grating, slight inclination for easy loading of the floor elements.

Specifications
  • Description:
    Sump pallet, PE, 250 l with PE grating, load capacity 1000 kg, weight 25,7 kg
  • Seq:
    10
  • W×D×H:
    1600×1200×180 mm
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Cross connector, short, for ground protection system
    Cross connector, short, for ground protection system
    CEMG10723
    115×115×35 mm
    Log in for availability
    1 * 1 ST - SFr. 21.80

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Connecting rail, short, for ground protection system
    Connecting rail, short, for ground protection system
    CEMG10722
    1540×15×15 mm
    Log in for availability
    1 * 1 ST - SFr. 35.80

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Access ramp, for ground protection system, load capacity 300 kg, weight 13 kg
    Access ramp, for ground protection system, load capacity 300 kg, weight 13 kg
    CEMG10714
    1260×800×200 mm
    Log in for availability
    1 * 1 ST - SFr. 370.00

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Connecting rail, long, for ground protection system
    Connecting rail, long, for ground protection system
    CEMG10721
    1140×15×15 mm
    Log in for availability
    1 * 1 ST - SFr. 28.60