Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Sump Pallets

Sump pallet, PE, 60 l, weight 3,0 kg, 800×600×165 mm
  CEMG10708
 :   CEMO
Sump Pallets
  CEMG10708
 :  CEMO
Sump Pallets
Sump Pallets

Sump pallet, PE, 60 l, weight 3,0 kg, 800×600×165 mm

 
SFr. 146.00
   
1 * 1 ST
 

These small container/laboratory trays can be combined to cover a complete storage area and also can be used as storage/filling station. Trays can be used with or without perforated plate.

  • Made of polyethylene
  • With general construction inspection approval (application for admission)
  • Stackable and provides space-saving storage/transport
  • Fully recyclable and maintenance friendly
  • Rugged, removable PE grating
  • Can be connected to form a surface protection system

Stable PE access ramp with an integrated fixing bolt for connection to the PE grating, slight inclination for easy loading of the floor elements.

 
  • Description:
    Sump pallet, PE, 60 l, weight 3,0 kg
  • Seq:
    05
  • W×D×H:
    800×600×165 mm
Accessories

 

  •  
    Description
    W×D×H
     
     
  • CEMG10723
    Cross connector, short, for ground protection system
    115×115×35 mm
       
    1 * 1 ST - SFr. 21.80

 

  •  
    Description
    W×D×H
     
     
  • CEMG10722
    Connecting rail, short, for ground protection system
    1540×15×15 mm
       
    1 * 1 ST - SFr. 35.80

 

  •  
    Description
    W×D×H
     
     
  • CEMG10714
    Access ramp, for ground protection system, load capacity 300 kg, weight 13 kg
    1260×800×200 mm
       
    1 * 1 ST - SFr. 370.00

 

  •  
    Description
    W×D×H
     
     
  • CEMG10721
    Connecting rail, long, for ground protection system
    1140×15×15 mm
       
    1 * 1 ST - SFr. 28.60

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.