Microplate reader, SpectraMax® L
Product Details & Documents
The SpectraMax® L microplate reader measures flash and glow luminescence assays in both 96- and 384-well plates.
- Detect dim samples with unsurpassed sensitivity
- Avoid saturation issues with 9 orders of dynamic range
- Enable robust performance with autorinse injectors
- Ensure compliance with IQ/OQ and FDA CFR 21 Part 11 compliance tools
- Easy set-up and analysis with industry-leading SoftMax Pro software
SpectraMax® L is upgradeable to include a barcode reader, programmable injectors, and dual-channel configurations for a 2-fold increase in medium throughput. The high sensitivity, low-noise photomultiplier tubes (PMTs), and variable-volume injector configuration make this reader ideal for fast kinetic assays requiring continuous monitoring during injection and measurement.
Dual injector option includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Dual injector option (auto aperture) includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
Two PMTs with automatic switching option enables wavelength-based measurements such as bioluminescence resonance energy transfer (BRET) assays and a 2-fold increase in throughput for traditional luminescence measurements. Wavelength-based measurements require purchase of appropriate filters.
Four injector option is suitable for SpectraMax® L readers with two PMTs and includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.
: For full details about the options available with this system please contact your local Avantor sales office.
: System includes SpectraMax® L luminometer for reading 96- and 384- well microplates with current version of SoftMax Pro software, AutoRange for >9 orders of dynamic range, Photon Counting for high sensitivity, landscape/portrait plate reading, incubator, user manual, USB interface cable, 1 year MDCare PhD Technical Support Plan and 1 year warranty covering parts and labour.
Specifications
- Description:Four injector option for SpectraMax® L readers with two PMTs
- Seq:10
Complementary Products
Product Information
- Catalog #DescriptionSerum formatSterilityPkRecommended working volumeColourMedia FormatPackedCulture areaSterileAvailabilityPrice
- MLDV5005331Loading tool for facilitating loading and unloading platesLog in for availability1 * 1 ST - SFr. 3’149.00
- S1810-500Foetal bovine serum, South American originSterile filtered15Log in for availability1 * 500 mL - SFr. 330.00
- 30028-L1Steady-Luc™ Firefly HTS assay kit (lyophilised)Log in for availability1 * 120 Tests - SFr. 180.00
- 30081-1Firefly and Renilla Luciferase Single Tube AssayLog in for availability1 * 150 Essai - SFr. 391.00
- 734-23271/bag96-well plates, flat bottom, TC-treated0,33 cm²+17ClearLog in for availability1 * 100 ST - SFr. 305.00