Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Microplate reader, SpectraMax® L | Avantor

Microplate reader, SpectraMax® L

Dual injector option for SpectraMax® L reader (auto aperture)
Katalognr. MLDV0310-5882
Anbieter:   Molecular Devices
Microplate reader, SpectraMax® L
Katalognr. MLDV0310-5882
Anbieter:  Molecular Devices
Microplate reader, SpectraMax® L
Microplate reader, SpectraMax® L

Dual injector option for SpectraMax® L reader (auto aperture)

Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents

The SpectraMax® L microplate reader measures flash and glow luminescence assays in both 96- and 384-well plates.

  • Detect dim samples with unsurpassed sensitivity
  • Avoid saturation issues with 9 orders of dynamic range
  • Enable robust performance with autorinse injectors
  • Ensure compliance with IQ/OQ and FDA CFR 21 Part 11 compliance tools
  • Easy set-up and analysis with industry-leading SoftMax Pro software

SpectraMax® L is upgradeable to include a barcode reader, programmable injectors, and dual-channel configurations for a 2-fold increase in medium throughput. The high sensitivity, low-noise photomultiplier tubes (PMTs), and variable-volume injector configuration make this reader ideal for fast kinetic assays requiring continuous monitoring during injection and measurement.

Dual injector option includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.

Dual injector option (auto aperture) includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.

Two PMTs with automatic switching option enables wavelength-based measurements such as bioluminescence resonance energy transfer (BRET) assays and a 2-fold increase in throughput for traditional luminescence measurements. Wavelength-based measurements require purchase of appropriate filters.

Four injector option is suitable for SpectraMax® L readers with two PMTs and includes two injectors for the measurement of single and dual flash assays in both 96- and 384-well microplates, Autorinse for the automatic recovery of reagent and rinsing of injector tubing at instrument shut down, and automatic disposal of wash and priming solutions.

Certifications: For full details about the options available with this system please contact your local Avantor sales office.

Delivery information: System includes SpectraMax® L luminometer for reading 96- and 384- well microplates with current version of SoftMax Pro software, AutoRange for >9 orders of dynamic range, Photon Counting for high sensitivity, landscape/portrait plate reading, incubator, user manual, USB interface cable, 1 year MDCare PhD Technical Support Plan and 1 year warranty covering parts and labour.

Technische Daten
  • Description:
    Dual injector option for SpectraMax® L reader (auto aperture)
  • Seq:
    08
Complementary Products

Produktinformationen

  • Katalognr.
    Description
    Serum format
    Sterility
    Pk
    Recommended working volume
    Colour
    Media Format
    Packed
    Culture area
    Sterile
    Verfügbarkeit
    Preis
  • MLDV0200-6017
    40-plate stacker magazines (pair)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 0.00

  • MLDV0200-6016
    20-plate stacker magazines (pair)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 0.00

  • MLDV0200-6187
    50-plate stacker magazines (pair)
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 0.00

  • MLDV5005331
    Loading tool for facilitating loading and unloading plates
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 3’149.00

  • S1810-500
    Foetal bovine serum, South American origin
    Sterile filtered
    15
    Einloggen für Verfügbarkeit
    1 * 500 mL - SFr. 330.00

  • MLDVSTAKMAX
    StakMax® microplate handling system
    Einloggen für Verfügbarkeit
    1 * 1 ST - SFr. 0.00

  • 30028-L1
    Steady-Luc™ Firefly HTS assay kit (lyophilised)
    Einloggen für Verfügbarkeit
    1 * 120 Tests - SFr. 180.00

  • 30081-1
    Firefly and Renilla Luciferase Single Tube Assay
    Einloggen für Verfügbarkeit
    1 * 150 Essai - SFr. 391.00

  • X0930-100
    Trypsin-EDTA 10X
    11
    Einloggen für Verfügbarkeit
    1 * 100 mL - SFr. 22.50

  • 734-2097
    200 µl
    06
    Transparent
    Einloggen für Verfügbarkeit
    1 * 50 ST - SFr. 271.00

  • L0135-500
    Ham's F12 with L-glutamine
    500 ml
    67
    Einloggen für Verfügbarkeit
    1 * 500 mL - SFr. 16.00

  • 735-0523
    Microlite™ 1+ strip, 1×12 per strip
    Einloggen für Verfügbarkeit
    1 * 320 ST - SFr. 252.00

  • 734-2327
    1/bag
    96-well plates, flat bottom, TC-treated
    0,33 cm²
    +
    17
    Clear
    Einloggen für Verfügbarkeit
    1 * 100 ST - SFr. 305.00

  • CORN3904BC
    20/bag
    +
    75 - 200 µl
    21
    360 µl
    Black
    Einloggen für Verfügbarkeit
    1 * 100 ST - SFr. 1’380.00