ContactUsLinkComponent
Jalas Clogs Occupational 5002 Menu Black, Regular Fit, PU-Coated Leather, Low Weight, oil resistant outsole, anti-static properties, adjustable heel strap, double shock absorp zones, ESD, Indoor Use 1 * 1 PAIR
Catalog # EJEN5002-42
Supplier:  EJENDALS
Jalas Clogs Occupational 5002 Menu Black, Regular Fit, PU-Coated Leather, Low Weight, oil resistant outsole, anti-static properties, adjustable heel strap, double shock absorp zones, ESD, Indoor Use 1 * 1 PAIR
Catalog # EJEN5002-42
Supplier:  EJENDALS