Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Erlenmeyer flask, PP, Corning®

Erlenmeyer flask, PP, Corning®

250 ml, Aseptic transfer cap, 20”
Catalog # CORN11410
   Low Manufacturing Impact
Supplier:   Corning
Erlenmeyer flask, PP, Corning®
Catalog # CORN11410
Supplier:  Corning
   Low Manufacturing Impact
Erlenmeyer flask, PP, Corning®
Erlenmeyer flask, PP, Corning®

250 ml, Aseptic transfer cap, 20”

Price
SFr. 930.00
Log in for availability
1 * 4 ST
Product Details & Documents

These closed system erlenmeyer flasks are ideal to reduce contamination risks and enhance the productivity of your shaker culture operations when adding or removing medium and cells.

  • Chemically resistant, sterile
  • Heat sealable flexible tubing
  • Male luer lock disconnect
  • Cap colour: Orange

Flask is made from optically clear polycarbonate and is preassembled with an aseptic transfer cap.

Certifications: Meets USP, Class VI requirements for plastic containers and closures.

Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904

Specifications
  • Closure type:
    Aseptic transfer cap
  • Capacity:
    250 ml
  • Seq:
    02
  • Tubing length:
    20”
  • Cat. No.:
    CORN11410
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..