ContactUsLinkComponent
Biological Indicatorological Indicator Spore Coupons for Monitoring Hydrogen Peroxide / Plasma Processes, Geobacillus stearothermoph ilus Cell Line 7953, Stainless Steel Coupons each 57 x 70 mm Individually Packaged in Tyvek®/Mylar® Pouches, 100 per 1 * 1 Each
Catalog # NAMSCPS-05
Supplier:  CROSSTEX
Biological Indicatorological Indicator Spore Coupons for Monitoring Hydrogen Peroxide / Plasma Processes, Geobacillus stearothermoph ilus Cell Line 7953, Stainless Steel Coupons each 57 x 70 mm Individually Packaged in Tyvek®/Mylar® Pouches, 100 per 1 * 1 Each
Catalog # NAMSCPS-05
Supplier:  CROSSTEX
Frequently Bought Together