ContactUsLinkComponent
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, Stain less Steel Disc each 6 mm in Diameter, Individually Packaged in Tyvek®/Tyvek® Pouches, 100 per box, 106 population le 1 * 1 Each
Catalog # NAMSTTN-06
Supplier:  CROSSTEX
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, Stain less Steel Disc each 6 mm in Diameter, Individually Packaged in Tyvek®/Tyvek® Pouches, 100 per box, 106 population le 1 * 1 Each
Catalog # NAMSTTN-06
Supplier:  CROSSTEX
Frequently Bought Together