ContactUsLinkComponent
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, Stain less Steel Discs each 6 mm in Diameter, Individually Packaged in Tyvek®/Mylar® Pouches, 100 per box, 106 population l 1 * 1 Each
Katalognr. NAMSSDN-06
Supplier:  CROSSTEX
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, Stain less Steel Discs each 6 mm in Diameter, Individually Packaged in Tyvek®/Mylar® Pouches, 100 per box, 106 population l 1 * 1 Each
Katalognr. NAMSSDN-06
Supplier:  CROSSTEX
Frequently Bought Together