ContactUsLinkComponent
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, 9 mm Glass Fiber Discs Individually Packaged in Tyvek/Mylar envelopes, 100 per box, 106 population level, ISO 11138 and USP 1 * 1 Each
Catalog # NAMSGFTN-06
Supplier:  CROSSTEX
Biological Indicatorological Indicators for Monitoring Hydrogen Peroxide/Plasma Processes, Bacillus atrophaeus Cell Line 9372, 9 mm Glass Fiber Discs Individually Packaged in Tyvek/Mylar envelopes, 100 per box, 106 population level, ISO 11138 and USP 1 * 1 Each
Catalog # NAMSGFTN-06
Supplier:  CROSSTEX
Frequently Bought Together