ContactUsLinkComponent
Biological Indicatorological Indicator Spore Strips for Monitoring Steam Processes, Geobacillus stearothermophilus Cell Line 7953, 6 mm x 30 mm Strips Individually Packaged in Glassine Envelopes, 100 strips per box, 102 population level, ISO 11138 a 1 * 100 items
Catalog # NAMSBS-102
Supplier:  CROSSTEX
Biological Indicatorological Indicator Spore Strips for Monitoring Steam Processes, Geobacillus stearothermophilus Cell Line 7953, 6 mm x 30 mm Strips Individually Packaged in Glassine Envelopes, 100 strips per box, 102 population level, ISO 11138 a 1 * 100 items
Catalog # NAMSBS-102
Supplier:  CROSSTEX
Frequently Bought Together