ZOBELL SOLUTION ORP CALIBRATOR 1 * 500 mL
Catalog # YSIA061322
Supplier: YSI ENVIRONMENTAL
Frequently Bought Together
VWR® Mortars with Pestles, Deep Form
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: PanReac AppliChem