ContactUsLinkComponent
HACCP digital pocket thermometer with blue colour coding, with labels also supplied in red, green, yellow and orange. BioCote® anti microbial protection, °C scale and auto switch off. Range -40°C to +125°C thermistor. 3.5mm jack connector. Supplied a 1 * 1 items
Catalog # COMAC12
Supplier:  COMARK
HACCP digital pocket thermometer with blue colour coding, with labels also supplied in red, green, yellow and orange. BioCote® anti microbial protection, °C scale and auto switch off. Range -40°C to +125°C thermistor. 3.5mm jack connector. Supplied a 1 * 1 items
Catalog # COMAC12
Supplier:  COMARK
Frequently Bought Together