Column packing support, Quickfit®
16 mm, 19/26 NS
BIBBEX13/12
PYREX®, borosilicate glass.
Designed to support Raschig rings within plain fractionating columns. Support is passed through socket of column and rests at the constriction above the cone.
- Cone size:19/26 NS
- Ø:16 mm
- Seq:01