2002-9050 15 ml HDPE Bottle only. Process: Particulate Cleaning & Gamma Irradiation Packaging: Single cleanroom bag each bottle. 72 per case. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certifi 1 * 72 items
Catalog # NALGCE-N7M15PI-EP
Supplier: Thermo Fisher Scientific
Frequently Bought Together