Orders LinkContactUsLinkComponent
Test kits, drop count tests
Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N)., 1 KIT
  HACH2437500
 :   Hach
Test kits, drop count tests
  HACH2437500
 :  Hach
 :  
Test kits, drop count tests
Test kits, drop count tests
Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N)., 1 KIT
 :  
SFr. 71.10
1 * 1 KIT
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with colour indicator
  • Reagents supplied with the kit
 : Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
  • Pk:
    1 KIT
  • Description:
    Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0,01 N).