ContactUsLinkComponent
2002-9025 8 mL HDPE Bottle only. Process: Particulate Cleaning & Gamma Irradiation. Packaging: Single cleanroom bag each bottle. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certificate of Proce 1 * 72 items
Catalog # NALGCE-N7M8PI-EP
2002-9025 8 mL HDPE Bottle only. Process: Particulate Cleaning & Gamma Irradiation. Packaging: Single cleanroom bag each bottle. Certification: C of A Endotoxin (Report Actual Results), C of A USP Particles, C of A USP Sterility, Certificate of Proce 1 * 72 items
Catalog # NALGCE-N7M8PI-EP