ContactUsLinkComponent
ChemiDoc-It 815 Cam/Dkrm Kit815 camera: 8.1 mp, 16-bit monochrome, digital camera and mount, with binning capabilities, cooling to - 57C from ambient, MOTORISED fixed 25mm, f/0.95 lens, close-up diopter, USB 2.0 enhanced card and cables, Extremely l 1 * 1 items
Catalog # UVPA97-0725-02
ChemiDoc-It 815 Cam/Dkrm Kit815 camera: 8.1 mp, 16-bit monochrome, digital camera and mount, with binning capabilities, cooling to - 57C from ambient, MOTORISED fixed 25mm, f/0.95 lens, close-up diopter, USB 2.0 enhanced card and cables, Extremely l 1 * 1 items
Catalog # UVPA97-0725-02
Frequently Bought Together