ContactUsLinkComponent
CF440 streptavidin conjugate is prepared from high quality streptavidin and CF440 (abs/em=440 nm/515 nm). CF440 is a superior al ternative to Alexa Fluor 430 owing to its exceptional photostability. null
  29066.
 :  Biotium
CF440 streptavidin conjugate is prepared from high quality streptavidin and CF440 (abs/em=440 nm/515 nm). CF440 is a superior al ternative to Alexa Fluor 430 owing to its exceptional photostability. null
  29066.
 :  Biotium
Frequently Bought Together