ContactUsLinkComponent
CF430 streptavidin conjugate is prepared from high quality streptavidin and CF430. It is a superior alternative to Pacific Green a nd BD Horizon V500 owing to its exceptional specific binding and photostability. It could be excited by violet laser 1 * 1 mg
Catalog # 29065.
Supplier:  Biotium
CF430 streptavidin conjugate is prepared from high quality streptavidin and CF430. It is a superior alternative to Pacific Green a nd BD Horizon V500 owing to its exceptional specific binding and photostability. It could be excited by violet laser 1 * 1 mg
Catalog # 29065.
Supplier:  Biotium
Frequently Bought Together