ContactUsLinkComponent
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: goat Form: lyophilized (IgG) Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855325
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: goat Form: lyophilized (IgG) Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855325
Supplier:  MP Biomedicals
Frequently Bought Together