ContactUsLinkComponent
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO HUMAN KAPPA LIGHT CHAIN (FREE BENCE - JONES PROTEIN). Anti-Human Host: goat Form: lyop hilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue secti 1 * 2 mL
Catalog # ICNA0855159
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED GOAT IgG FRACTION TO HUMAN KAPPA LIGHT CHAIN (FREE BENCE - JONES PROTEIN). Anti-Human Host: goat Form: lyop hilized (IgG) Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue secti 1 * 2 mL
Catalog # ICNA0855159
Supplier:  MP Biomedicals
Frequently Bought Together