ContactUsLinkComponent
PURIFIED MOUSE MYELOMA PROTEIN IgG3 (KAPPA) FLOPC 21. These myeloma proteins are purified from ascites by a combination of salt precipitation, ion exchange and bioaffinity chromatography. 1 * 1 mg
  ICNA0850332
PURIFIED MOUSE MYELOMA PROTEIN IgG3 (KAPPA) FLOPC 21. These myeloma proteins are purified from ascites by a combination of salt precipitation, ion exchange and bioaffinity chromatography. 1 * 1 mg
  ICNA0850332
Frequently Bought Together